2-(3-Chlorophenoxy)nicotinamide

Supplier: Matrix Scientific

039739-500MG
101848-088EA 216.92 USD
101848-088
2-(3-Chlorophenoxy)nicotinamide
2-(3-Chlorophenoxy)nicotinamide

Hazard: Irritant

Formula: C₁₂H₉ClN₂O₂
MDL Number: MFCD02325548
CAS Number: 214758-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR