(1-Methyl-1H-pyrazol-3-yl)acetic acid
Supplier: Matrix Scientific
039737-500MG
101848-084EA
476.92
USD
101848-084
(1-Methyl-1H-pyrazol-3-yl)acetic acid
(1-Methyl-1H-pyrazol-3-yl)acetic acid
Hazard: Irritant
Formula:
C₆H₈N₂O₂ MW: 140.14 g/mol |
MDL Number:
MFCD12028051 CAS Number: 1177318-00-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...