2-Methyl-6-nitropyrazolo[1,5-a]pyrimidine

Supplier: Matrix Scientific

039735-500MG
101848-080EA 290.77 USD
101848-080
2-Methyl-6-nitropyrazolo[1,5-a]pyrimidine
2-Methyl-6-nitropyrazolo[1,5-a]pyrimidine

Hazard: Irritant

Formula: C₇H₆N₄O₂
MDL Number: MFCD00296942
CAS Number: 80772-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR