Methyl [(5-methyl-4-nitro-1H-pyrazol-3-yl)thio]acetate

Supplier: Matrix Scientific

039734-500MG
101848-078EA 217.35 USD
101848-078
Methyl [(5-methyl-4-nitro-1H-pyrazol-3-yl)thio]acetate
Methyl [(5-methyl-4-nitro-1H-pyrazol-3-yl)thio]acetate

Hazard: Irritant

Formula: C₇H₉N₃O₄S
MDL Number: MFCD12028049
CAS Number: 1240284-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR