2-Cyano-N-nonylacetamide

Supplier: Matrix Scientific

039723-500MG
101848-056EA 146.15 USD
101848-056
2-Cyano-N-nonylacetamide
2-Cyano-N-nonylacetamide

Hazard: Irritant

Formula: C₁₂H₂₂N₂O
MDL Number: MFCD03377520
CAS Number: 52493-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR