N-[4-(Aminosulfonyl)benzyl]-2-cyanoacetamide

Supplier: Matrix Scientific

039721-500MG
101848-052EA 109.25 USD
101848-052
N-[4-(Aminosulfonyl)benzyl]-2-cyanoacetamide
N-[4-(Aminosulfonyl)benzyl]-2-cyanoacetamide

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₃S
MDL Number: MFCD03071693
CAS Number: 32933-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR