Ethyl-4-(4-bromophenyl)-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

Supplier: Matrix Scientific

039720-500MG
101848-050EA 387.69 USD
101848-050
Ethyl-4-(4-bromophenyl)-2-[(cyanoacetyl)amino]thiophene-3-carboxylate
Ethyl-4-(4-bromophenyl)-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₃BrN₂O₃S
MDL Number: MFCD03392237
CAS Number: 546064-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR