1-(Cyanoacetyl)piperidine

Supplier: Matrix Scientific

039719-500MG
101848-048EA 109.25 USD
101848-048
1-(Cyanoacetyl)piperidine
1-(Cyanoacetyl)piperidine

Hazard: Irritant

Formula: C₈H₁₂N₂O
MDL Number: MFCD00006487
CAS Number: 15029-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR