Methyl-2-[(cyanoacetyl)amino]-4-(4-fluorophenyl)thiophene-3-carboxylate

Supplier: Matrix Scientific

039718-500MG
101848-046EA 289.8 USD
101848-046
Methyl-2-[(cyanoacetyl)amino]-4-(4-fluorophenyl)thiophene-3-carboxylate
Methyl-2-[(cyanoacetyl)amino]-4-(4-fluorophenyl)thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₁FN₂O₃S
MDL Number: MFCD03396396
CAS Number: 546116-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR