2-Cyano-N-(3-cyano-4,5,6,7,8,9-hexahydrocycloocta[b]thien-2-yl)acetamide

Supplier: Matrix Scientific

039717-500MG
101848-044EA 290.77 USD
101848-044
2-Cyano-N-(3-cyano-4,5,6,7,8,9-hexahydrocycloocta[b]thien-2-yl)acetamide
2-Cyano-N-(3-cyano-4,5,6,7,8,9-hexahydrocycloocta[b]thien-2-yl)acetamide

Hazard: Irritant

Formula: C₁₄H₁₅N₃OS
MDL Number: MFCD04141307
CAS Number: 725222-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR