Propyl-2-[(cyanoacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Supplier: Matrix Scientific

039715-500MG
101848-040EA 290.77 USD
101848-040
Propyl-2-[(cyanoacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Propyl-2-[(cyanoacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₃S
MW: 320.41 g/mol
MDL Number: MFCD01337323
CAS Number: 574717-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR