N-(1-Bicyclo[2.2.1]hept-2-ylethyl)-2-cyanoacetamide

Supplier: Matrix Scientific

039712-500MG
101848-034EA 217.35 USD
101848-034
N-(1-Bicyclo[2.2.1]hept-2-ylethyl)-2-cyanoacetamide
N-(1-Bicyclo[2.2.1]hept-2-ylethyl)-2-cyanoacetamide

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD01345322
CAS Number: 1023832-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR