3-[4-(2,3-Dimethylphenyl)piperazin-1-yl]-3-oxopropanenitrile

Supplier: Matrix Scientific

039711-500MG
101848-032EA 109.25 USD
101848-032
3-[4-(2,3-Dimethylphenyl)piperazin-1-yl]-3-oxopropanenitrile
3-[4-(2,3-Dimethylphenyl)piperazin-1-yl]-3-oxopropanenitrile

Hazard: Irritant

Formula: C₁₅H₁₉N₃O
MDL Number: MFCD02174244
CAS Number: 543717-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR