Ethyl-2-[(cyanoacetyl)amino]-4,5-dimethylthiophene-3-carboxylate

Supplier: Matrix Scientific

039708-500MG
101848-026EA 289.8 USD
101848-026
Ethyl-2-[(cyanoacetyl)amino]-4,5-dimethylthiophene-3-carboxylate
Ethyl-2-[(cyanoacetyl)amino]-4,5-dimethylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₃S
MDL Number: MFCD02323196
CAS Number: 405924-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR