2-Cyano-N-[2-(4-methoxyphenyl)ethyl]acetamide

Supplier: Matrix Scientific

039707-500MG
101848-024EA 146.15 USD
101848-024
2-Cyano-N-[2-(4-methoxyphenyl)ethyl]acetamide
2-Cyano-N-[2-(4-methoxyphenyl)ethyl]acetamide

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₂
MDL Number: MFCD01341616
CAS Number: 100134-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR