Ethyl-2-[(cyanoacetyl)amino]-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
Supplier: Matrix Scientific
039706-500MG
101848-022EA
290.77
USD
101848-022
Ethyl-2-[(cyanoacetyl)amino]-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
Ethyl-2-[(cyanoacetyl)amino]-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
Hazard: Irritant
Formula:
C₁₅H₁₈N₂O₃S |
MDL Number:
MFCD04060005 CAS Number: 669753-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...