2-Cyano-N-(2-nitrophenyl)acetamide

Supplier: Matrix Scientific

039705-500MG
101848-020EA 146.15 USD
101848-020
2-Cyano-N-(2-nitrophenyl)acetamide
2-Cyano-N-(2-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₉H₇N₃O₃
MDL Number: MFCD01910047
CAS Number: 65372-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR