2-Cyano-N-(4-nitrophenyl)acetamide

Supplier: Matrix Scientific

039704-500MG
101848-018EA 109.25 USD
101848-018
2-Cyano-N-(4-nitrophenyl)acetamide
2-Cyano-N-(4-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₉H₇N₃O₃
MDL Number: MFCD00828749
CAS Number: 22208-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR