N-(1,3-Benzodioxol-5-ylmethyl)-2-cyanoacetamide

Supplier: Matrix Scientific

039703-500MG
101848-016EA 109.25 USD
101848-016
N-(1,3-Benzodioxol-5-ylmethyl)-2-cyanoacetamide
N-(1,3-Benzodioxol-5-ylmethyl)-2-cyanoacetamide

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₃
MDL Number: MFCD01345888
CAS Number: 341953-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR