N-(4-Butylphenyl)-2-cyanoacetamide

Supplier: Matrix Scientific

039700-500MG
101848-010EA 146.15 USD
101848-010
N-(4-Butylphenyl)-2-cyanoacetamide
N-(4-Butylphenyl)-2-cyanoacetamide

Hazard: Irritant

Formula: C₁₃H₁₆N₂O
MDL Number: MFCD01356438
CAS Number: 54153-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR