2-[(Cyanoacetyl)amino]-5-isopropylthiophene-3-carboxamide

Supplier: Matrix Scientific

039699-500MG
101848-008EA 289.8 USD
101848-008
2-[(Cyanoacetyl)amino]-5-isopropylthiophene-3-carboxamide
2-[(Cyanoacetyl)amino]-5-isopropylthiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₂S
MDL Number: MFCD04054711
CAS Number: 667414-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR