2-Cyano-N-[3-cyano-6-(1,1-dimethylpropyl)-4,5,6,7-tetrahydro-1-benzothien-2-yl]acetamide

Supplier: Matrix Scientific

039698-500MG
101848-006EA 290.77 USD
101848-006
2-Cyano-N-[3-cyano-6-(1,1-dimethylpropyl)-4,5,6,7-tetrahydro-1-benzothien-2-yl]acetamide
2-Cyano-N-[3-cyano-6-(1,1-dimethylpropyl)-4,5,6,7-tetrahydro-1-benzothien-2-yl]acetamide

Hazard: Irritant

Formula: C₁₇H₂₁N₃OS
MDL Number: MFCD04054638
CAS Number: 667412-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR