2-Cyano-N-mesitylacetamide

Supplier: Matrix Scientific

039697-500MG
101848-004EA 146.15 USD
101848-004
2-Cyano-N-mesitylacetamide
2-Cyano-N-mesitylacetamide

Hazard: Irritant

Formula: C₁₂H₁₄N₂O
MDL Number: MFCD00278154
CAS Number: 24578-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR