4-Cyano-5-[(cyanoacetyl)amino]-N,N-3-trimethylthiophene-2-carboxamide

Supplier: Matrix Scientific

039695-500MG
101848-000EA 289.8 USD
101848-000
4-Cyano-5-[(cyanoacetyl)amino]-N,N-3-trimethylthiophene-2-carboxamide
4-Cyano-5-[(cyanoacetyl)amino]-N,N-3-trimethylthiophene-2-carboxamide

Hazard: Irritant

Formula: C₁₂H₁₂N₄O₂S
MDL Number: MFCD04054576
CAS Number: 667412-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR