N-[4-(Aminosulfonyl)phenyl]-2-cyanoacetamide

Supplier: Matrix Scientific

039693-500MG
101847-996EA 193.85 USD
101847-996
N-[4-(Aminosulfonyl)phenyl]-2-cyanoacetamide
N-[4-(Aminosulfonyl)phenyl]-2-cyanoacetamide

Hazard: Irritant

Formula: C₉H₉N₃O₃S
MDL Number: MFCD01115985
CAS Number: 32933-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR