Ethyl-4-(4-chlorophenyl)-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

Supplier: Matrix Scientific

039691-500MG
101847-992EA 289.8 USD
101847-992
Ethyl-4-(4-chlorophenyl)-2-[(cyanoacetyl)amino]thiophene-3-carboxylate
Ethyl-4-(4-chlorophenyl)-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₃ClN₂O₃S
MDL Number: MFCD03398011
CAS Number: 547706-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR