2-Cyano-N-(pyridin-2-ylmethyl)acetamide

Supplier: Matrix Scientific

039687-500MG
101847-984EA 146.15 USD
101847-984
2-Cyano-N-(pyridin-2-ylmethyl)acetamide
2-Cyano-N-(pyridin-2-ylmethyl)acetamide

Hazard: Irritant

Formula: C₉H₉N₃O
MDL Number: MFCD00828756
CAS Number: 84951-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR