Ethyl-2-[(cyanoacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Supplier: Matrix Scientific

039685-500MG
101847-980EA 290.77 USD
101847-980
Ethyl-2-[(cyanoacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Ethyl-2-[(cyanoacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₃S
MDL Number: MFCD01357697
CAS Number: 670233-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR