N-[2-Chloro-5-(trifluoromethyl)phenyl]-2-cyanoacetamide

Supplier: Matrix Scientific

039684-500MG
101847-978EA 146.15 USD
101847-978
N-[2-Chloro-5-(trifluoromethyl)phenyl]-2-cyanoacetamide
N-[2-Chloro-5-(trifluoromethyl)phenyl]-2-cyanoacetamide

Hazard: Irritant

Formula: C₁₀H₆ClF₃N₂O
MDL Number: MFCD00116912
CAS Number: 63034-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR