N-1,3-Benzodioxol-5-yl-2-cyanoacetamide

Supplier: Matrix Scientific

039683-500MG
101847-976EA 146.15 USD
101847-976
N-1,3-Benzodioxol-5-yl-2-cyanoacetamide
N-1,3-Benzodioxol-5-yl-2-cyanoacetamide

Hazard: Irritant

Formula: C₁₀H₈N₂O₃
MDL Number: MFCD03152807
CAS Number: 142555-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR