Ethyl-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

Supplier: Matrix Scientific

039679-500MG
101847-968EA 289.8 USD
101847-968
Ethyl-2-[(cyanoacetyl)amino]thiophene-3-carboxylate
Ethyl-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₃S
MDL Number: MFCD03900488
CAS Number: 667436-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR