Ethyl-2-[(cyanoacetyl)amino]-5-ethylthiophene-3-carboxylate

Supplier: Matrix Scientific

039678-500MG
101847-966EA 387.69 USD
101847-966
Ethyl-2-[(cyanoacetyl)amino]-5-ethylthiophene-3-carboxylate
Ethyl-2-[(cyanoacetyl)amino]-5-ethylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₃S
MDL Number: MFCD00122066
CAS Number: 667435-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR