Methyl-2-[(cyanoacetyl)amino]-5-ethylthiophene-3-carboxylate

Supplier: Matrix Scientific

039677-500MG
101847-964EA 241.5 USD
101847-964
Methyl-2-[(cyanoacetyl)amino]-5-ethylthiophene-3-carboxylate
Methyl-2-[(cyanoacetyl)amino]-5-ethylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃S
MDL Number: MFCD03900443
CAS Number: 667435-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR