2-Cyano-N-(4-isopropylphenyl)acetamide

Supplier: Matrix Scientific

039670-500MG
101847-950EA 146.15 USD
101847-950
2-Cyano-N-(4-isopropylphenyl)acetamide
2-Cyano-N-(4-isopropylphenyl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₄N₂O
MDL Number: MFCD01350113
CAS Number: 24522-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR