2-Cyano-N-(2,5-dimethylphenyl)acetamide
Supplier: Matrix Scientific
039668-500MG
101847-946EA
146.15
USD
101847-946
2-Cyano-N-(2,5-dimethylphenyl)acetamide
2-Cyano-N-(2,5-dimethylphenyl)acetamide
Hazard: Irritant
Formula:
C₁₁H₁₂N₂O |
MDL Number:
MFCD01429817 CAS Number: 87165-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...