2-Cyano-N-(3,5-dimethylphenyl)acetamide

Supplier: Matrix Scientific

039665-500MG
101847-940EA 146.15 USD
101847-940
2-Cyano-N-(3,5-dimethylphenyl)acetamide
2-Cyano-N-(3,5-dimethylphenyl)acetamide

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MDL Number: MFCD02194059
CAS Number: 340232-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR