Methyl-2-[(cyanoacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Supplier: Matrix Scientific

039662-500MG
101847-934EA 290.77 USD
101847-934
Methyl-2-[(cyanoacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Methyl-2-[(cyanoacetyl)amino]-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₃S
MDL Number: MFCD03946153
CAS Number: 590358-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR