3-(4-Benzylpiperidin-1-yl)-3-oxopropanenitrile
Supplier: Matrix Scientific
039661-500MG
101847-932EA
146.15
USD
101847-932
3-(4-Benzylpiperidin-1-yl)-3-oxopropanenitrile
3-(4-Benzylpiperidin-1-yl)-3-oxopropanenitrile
Hazard: Irritant
Formula:
C₁₅H₁₈N₂O |
MDL Number:
MFCD01353601 CAS Number: 551907-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...