Methyl-2-[(cyanoacetyl)amino]-4,5,6,7,8,9-hexahydrocycloocta[b]thiophene-3-carboxylate

Supplier: Matrix Scientific

039659-500MG
101847-928EA 290.77 USD
101847-928
Methyl-2-[(cyanoacetyl)amino]-4,5,6,7,8,9-hexahydrocycloocta[b]thiophene-3-carboxylate
Methyl-2-[(cyanoacetyl)amino]-4,5,6,7,8,9-hexahydrocycloocta[b]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₃S
MDL Number: MFCD01356585
CAS Number: 666728-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR