Ethyl-2-[(cyanoacetyl)amino]-4-ethyl-5-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

039658-500MG
101847-926EA 387.69 USD
101847-926
Ethyl-2-[(cyanoacetyl)amino]-4-ethyl-5-methylthiophene-3-carboxylate
Ethyl-2-[(cyanoacetyl)amino]-4-ethyl-5-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₃S
MDL Number: MFCD01346299
CAS Number: 549542-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR