Methyl-2-[(cyanoacetyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Supplier: Matrix Scientific

039657-500MG
101847-924EA 290.77 USD
101847-924
Methyl-2-[(cyanoacetyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Methyl-2-[(cyanoacetyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₃S
MDL Number: MFCD03378029
CAS Number: 544440-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR