Methyl-2-[(cyanoacetyl)amino]-4,5-dimethylthiophene-3-carboxylate

Supplier: Matrix Scientific

039656-500MG
101847-922EA 387.69 USD
101847-922
Methyl-2-[(cyanoacetyl)amino]-4,5-dimethylthiophene-3-carboxylate
Methyl-2-[(cyanoacetyl)amino]-4,5-dimethylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃S
MDL Number: MFCD03387850
CAS Number: 545347-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR