2-Cyano-N-(4-phenyl-1,3-thiazol-2-yl)acetamide

Supplier: Matrix Scientific

039655-500MG
101847-920EA 290.77 USD
101847-920
2-Cyano-N-(4-phenyl-1,3-thiazol-2-yl)acetamide
2-Cyano-N-(4-phenyl-1,3-thiazol-2-yl)acetamide

Hazard: Irritant

Formula: C₁₂H₉N₃OS
MDL Number: MFCD02251253
CAS Number: 31557-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR