3-[4-(1,3-Benzodioxol-5-ylmethyl)piperazin-1-yl]-3-oxopropanenitrile

Supplier: Matrix Scientific

039650-500MG
101847-910EA 146.15 USD
101847-910
3-[4-(1,3-Benzodioxol-5-ylmethyl)piperazin-1-yl]-3-oxopropanenitrile
3-[4-(1,3-Benzodioxol-5-ylmethyl)piperazin-1-yl]-3-oxopropanenitrile

Hazard: Irritant

Formula: C₁₅H₁₇N₃O₃
MDL Number: MFCD03396393
CAS Number: 546116-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR