Methyl-4-(4-bromophenyl)-2-[(cyanoacetyl)amino]thiophene-3-carboxylate
Supplier: Matrix Scientific
039649-500MG
101847-908EA
387.69
USD
101847-908
Methyl-4-(4-bromophenyl)-2-[(cyanoacetyl)amino]thiophene-3-carboxylate
Methyl-4-(4-bromophenyl)-2-[(cyanoacetyl)amino]thiophene-3-carboxylate
Hazard: Irritant
Formula:
C₁₅H₁₁BrN₂O₃S |
MDL Number:
MFCD03390676 CAS Number: 545385-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...