Isopropyl-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

Supplier: Matrix Scientific

039647-500MG
101847-904EA 387.69 USD
101847-904
Isopropyl-2-[(cyanoacetyl)amino]thiophene-3-carboxylate
Isopropyl-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃S
MDL Number: MFCD03945520
CAS Number: 590351-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR