Methyl-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

Supplier: Matrix Scientific

039646-500MG
101847-902EA 387.69 USD
101847-902
Methyl-2-[(cyanoacetyl)amino]thiophene-3-carboxylate
Methyl-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₉H₈N₂O₃S
MDL Number: MFCD03945477
CAS Number: 590351-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR