N-(4-Acetylphenyl)-2-cyanoacetamide

Supplier: Matrix Scientific

039644-500MG
101847-898EA 146.15 USD
101847-898
N-(4-Acetylphenyl)-2-cyanoacetamide
N-(4-Acetylphenyl)-2-cyanoacetamide

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MDL Number: MFCD03383082
CAS Number: 219618-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR