Methyl-2-[(cyanoacetyl)amino]-5-phenylthiophene-3-carboxylate

Supplier: Matrix Scientific

039643-500MG
101847-896EA 387.69 USD
101847-896
Methyl-2-[(cyanoacetyl)amino]-5-phenylthiophene-3-carboxylate
Methyl-2-[(cyanoacetyl)amino]-5-phenylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₂N₂O₃S
MDL Number: MFCD03759463
CAS Number: 515872-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR