Ethyl-2-[(cyanoacetyl)amino]-4-phenylthiophene-3-carboxylate
Supplier: Matrix Scientific
039642-500MG
101847-894EA
387.69
USD
101847-894
Ethyl-2-[(cyanoacetyl)amino]-4-phenylthiophene-3-carboxylate
Ethyl-2-[(cyanoacetyl)amino]-4-phenylthiophene-3-carboxylate
Hazard: Irritant
Formula:
C₁₆H₁₄N₂O₃S |
MDL Number:
MFCD03394771 CAS Number: 546090-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...